Orders LinkContactUsLinkComponent
Osmometer Calibration Standards
Osmometer Calibration Standards
Certificates
Osmometer Calibration Standards
Osmometer Calibration Standards
12 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Omolality calibration standards for clinical and research osmometers.
  • Includes Certificate of Analysis
  • For IVD and research use
  • Compatible with all Osmometers
  • 10×1 ml ampules (ideal volume for unit calibration)
  • Sturdy packaging
  • Two year shelf life
Four E's is a global manufacturer of clinical and research equipment and reagents. These IVD grade Osmometer Calibration Standards are for calibrating Osmometers, they are suitable for use with the Four E's Osmometer or any other Osmometers.
 : CoA and Accuracy statement.
 : Ampules are packaged in a cardboard box.
 : Do not shake prior to use, only swirl.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 76670-496
    Osmometer calibration standard 0 mOsm/kg
    Osmometer calibration standard 0 mOsm/kg
    10×1 ml ampule
    Log in for availability
    Log in for availability
    Box of 10 (1ml)
    Box of 10 (1ml) - $198.92
    $198.92
    Box of 10 (1ml)
       $198.92
    Supplier Number: 3MA0004ES
  • 76670-500
    Osmometer calibration standard 50 mOsm/kg
    Osmometer calibration standard 50 mOsm/kg
    10×1 ml ampule
    Log in for availability
    Log in for availability
    Box of 10 (1ml)
    Box of 10 (1ml) - $198.92
    $198.92
    Box of 10 (1ml)
       $198.92
    Supplier Number: 3MA0054ES
  • 76670-482
    Osmometer calibration standard 100 mOsm/kg
    Osmometer calibration standard 100 mOsm/kg
    10×1 ml ampule
    Log in for availability
    Log in for availability
    Box of 10 (1ml)
    Box of 10 (1ml) - $198.92
    $198.92
    Box of 10 (1ml)
       $198.92
    Supplier Number: 3LA0114ES
  • 76670-502
    Osmometer calibration standard 200 mOsm/kg
    Osmometer calibration standard 200 mOsm/kg
    10×1 ml ampule
    Discontinued
    Supplier Number: -
  • 76670-506
    Osmometer calibration standard 290 mOsm/kg
    Osmometer calibration standard 290 mOsm/kg
    10×1 ml ampule
    Log in for availability
    Log in for availability
    Box of 10 (1ml)
    Box of 10 (1ml) - $198.92
    $198.92
    Box of 10 (1ml)
       $198.92
    Supplier Number: 3MA0294ES
  • 76670-498
    Osmometer calibration standard 300 mOsm/kg
    Osmometer calibration standard 300 mOsm/kg
    10×1 ml ampule
    Log in for availability
    Log in for availability
    Box of 10 (1ml)
    Box of 10 (1ml) - $198.92
    $198.92
    Box of 10 (1ml)
       $198.92
    Supplier Number: 3MA0034ES
  • 76670-508
    Osmometer calibration standard 400 mOsm/kg
    Osmometer calibration standard 400 mOsm/kg
    10×1 ml ampule
    Log in for availability
    Log in for availability
    Box of 10 (1ml)
    Box of 10 (1ml) - $198.92
    $198.92
    Box of 10 (1ml)
       $198.92
    Supplier Number: 3MA0404ES
  • 76670-486
    Osmometer calibration standard 500 mOsm/kg
    Osmometer calibration standard 500 mOsm/kg
    10×1 ml ampule
    Log in for availability
    Log in for availability
    Box of 10 (1ml)
    Box of 10 (1ml) - $198.92
    $198.92
    Box of 10 (1ml)
       $198.92
    Supplier Number: 3LA0514ES
  • 76670-510
    Osmometer calibration standard 850 mOsm/kg
    Osmometer calibration standard 850 mOsm/kg
    10×1 ml ampule
    Log in for availability
    Log in for availability
    Box of 10 (1ml)
    Box of 10 (1ml) - $198.92
    $198.92
    Box of 10 (1ml)
       $198.92
    Supplier Number: 3MA0854ES
  • 76670-490
    Osmometer calibration standard 900 mOsm/kg
    Osmometer calibration standard 900 mOsm/kg
    10×1 ml ampule
    Log in for availability
    Log in for availability
    Box of 10 (1ml)
    Box of 10 (1ml) - $198.92
    $198.92
    Box of 10 (1ml)
       $198.92
    Supplier Number: 3LA0914ES
  • 76670-512
    Osmometer calibration standard 1000 mOsm/kg
    Osmometer calibration standard 1000 mOsm/kg
    10×1 ml ampule
    Discontinued
    Supplier Number: -
  • 76670-492
    Osmometer calibration standard 1500 mOsm/kg
    Osmometer calibration standard 1500 mOsm/kg
    10×1 ml ampule
    Log in for availability
    Log in for availability
    Box of 10 (1ml)
    Box of 10 (1ml) - $198.92
    $198.92
    Box of 10 (1ml)
       $198.92
    Supplier Number: 3LA1514ES
  • 76670-514
    Osmometer calibration standard 2000 mOsm/kg
    Osmometer calibration standard 2000 mOsm/kg
    10×1 ml ampule
    Log in for availability
    Log in for availability
    Box of 10 (1ml)
    Box of 10 (1ml) - $198.92
    $198.92
    Box of 10 (1ml)
       $198.92
    Supplier Number: 3MA2004ES
  • 76670-494
    Osmometer calibration standard 3000 mOsm/kg
    Osmometer calibration standard 3000 mOsm/kg
    10×1 ml ampule
    Discontinued
    Supplier Number: -
  • 76670-516
    Osmometer calibration standard 4000 mOsm/kg
    Osmometer calibration standard 4000 mOsm/kg
    10×1 ml ampule
    Discontinued
    Supplier Number: -
  • 76670-504
    Osmometer calibration standard, three level: 240/280/320 mOsm/kg
    Osmometer calibration standard, three level: 240/280/320 mOsm/kg
    9×1 ml ampule
    Discontinued
    Supplier Number: -
  • 76670-488
    Osmometer calibration standard, two level: 300/800 mOsm/kg
    Osmometer calibration standard, two level: 300/800 mOsm/kg
    10×1 ml ampule
    Discontinued
    Supplier Number: -
  • 76670-484
    Osmometer calibration standards linearity set: 100 - 2000 mOsm
    Osmometer calibration standards linearity set: 100 - 2000 mOsm
    10×1 ml ampule
    Log in for availability
    Log in for availability
    Box of 10 (1ml)
    Box of 10 (1ml) - $284.15
    $284.15
    Box of 10 (1ml)
       $284.15
    Supplier Number: 3LA0284ES