Ultra Sensitivity NGS Kit
76645-206
- Description:Ultra sensitivity NGS kit
- Cat. No.:76645-206
- Details:Femto pulse system kit for the quantitative and qualitative analysis of NGS libraries and intermediates from 100 - 6000 bp at femtogram-level concentrations, includes reagents necessary for running 275 samples
NGS library quality control for low concentration NGS libraries and DNA smears, including cfDNA.
- Enhanced detection - Visualization of DNA smears down to 5 pg/µl, DNA fragments down to 50 fg/µl enabling conservation of samples for downstream NGS library sequencing
- Sizing accuracy - Reliable sizing from 100 to 6000 bp, making the kit ideal for applications such as short-read NGS library QC, single cell genomics, and cfDNA
- Low sample input - A sample input of only 2 µl at very small concentrations minimizes the sample needed for NGS library QC, allowing the conservation of precious sample volumes for downstream sequencing analysis
- Smear analysis - ProSize data analysis software is used to calculate the precise molarity of an NGS library, leading to more efficient flow cell loading and optimal sequencing data
- Reliable quantification - Accurate and precise quantification of DNA smears up to 250 pg/µl
The ultra sensitivity NGS kit for the Agilent Femto pulse system is used for NGS library QC, providing sizing and quantification for DNA smears and fragments into the femtogram range. Detection of DNA smears as low as 5 pg/µl and quantification through 250 pg/µl makes the ultra-sensitivity NGS kit ideal for quality control of low concentration NGS libraries. Other genomics applications include single cell genomics, PCR-free NGS libraries, and cfDNA QC.
: For research use only. Not for use in diagnostic procedures.