Order Entry
Puerto Rico
ContactUsLinkComponent
Chloroform-[D₁] (99.8% D) + 0.03% (v/v) TMS, AcroSeal™ for NMR spectroscopy
Toxic
Health
Chloroform-[D₁] (99.8% D) + 0.03% (v/v) TMS, AcroSeal™ for NMR spectroscopy
Catalog # TS42696-1000
CAS Number:  
Toxic
Health
Chloroform-[D₁] (99.8% D) + 0.03% (v/v) TMS, AcroSeal™ for NMR spectroscopy
Catalog # TS42696-1000
Supplier Number:  42696-1000
CAS Number:  

Specifications

  • Size:
    100 ml
  • Cat. No.:
    TS42696-1000

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.02 %
  • D-Enrichment:
    ≥99.75 %
  • TMS:
    0.028 to 0.032 v/v%

Specification Test Results

About this item

 :
 :120.38 g/mol
Boiling Pt:60.8 °C (1013 hPa)
Melting Pt:–64 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-742-4
UN:1888
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:865-49-6
Formula:CDCl₃
MDL Number:MFCD00000827