Order Entry
Puerto Rico
ContactUsLinkComponent
Acetonitrile-[D₃] 99.8 atom % D for NMR spectroscopy
Flammable
Irritant
Acetonitrile-[D₃] 99.8 atom % D for NMR spectroscopy
Catalog # TS32066-0075
CAS Number:  
undefined
Flammable
Irritant
Acetonitrile-[D₃] 99.8 atom % D for NMR spectroscopy
Catalog # TS32066-0075
Supplier Number:  32066-0075
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Size:
    7.5 ml
  • Cat. No.:
    TS32066-0075

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.03 % (Coulometric)
  • D-Enrichment:
    ≥99.75 %

Specification Test Results

About this item

 :
 :44.03 g/mol
Boiling Pt:80.7 °C (1013 hPa)
Melting Pt:–46 °C
Density:0.844 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:218-616-5
UN:1648
ADR:3,II
 :GHS02
 :GHS07
CAS Number:2206-26-0
Formula:D₃CCN
MDL Number:MFCD00001881