CIMac PrimaS™ 0.1 ml Analytical Column, BIA Separations
Catalog # MSPP-110102113
Supplier: BIA SEPARATIONS INC. MS
Specifications
- Max. Pressure:15 MPa (2175 psi)
- Capacity:0.1 ml
- Description:CIMac™ r-Protein L analytical column (Recombinant Protein L) (1.3 µm channels)
- pH Range:Working range 2 - 13, cleaning in place 1 - 14
- Temperature Limit:4...40 °C
- Cat. No.:MSPP-110102113
Specifications
About this item
CIMac™ Analytical Monoliths are high performance chromatography devices for rapid highresolution fractionation of complex biological samples.
- Support matrix: Poly(glycidyl methacrylate -co- ethylene dimethacrylate)
- Monolith dimensions: Diameter: 5.2 mm; length: 4.95 mm; bed volume (CV): 0.1 ml
- Connector: 10 - 32 UNF coned port, ¹/₁₆" ext.Ø tubing connection
- Operating flow rates: 0.2-3 ml/min (1-15 cm/min; 2-30 CV/min)
- Operating temperature: 4 °C (39 °F) to 40 °C (104 °F)
- Chemical stability: All commonly used aqueous buffers, 8 M urea, 6 M guanidine hydrochloride and 20% ethanol solution.
- Storage conditions: 2 °C (36 °F) to 25 °C (77 °F); 20% ethanol
The stationary phase is polymerized as a monolith with homogeneous channel size and surface chemistry. Each unit is mounted in a precision engineered stainless steel housing to allow easy connection to any HPLC system.
PrimaS is a multimodal ligand which exploits a combination of anion exchange and hydrogen bonding to achieve unique selectivity. The following information is provided to ensure proper product care and optimal product performance.