About this item
FabRICATOR Z Fab2 Kit consists of spin columns of FabRICATOR Z Immobilized for antibody digestion, and spin columns of CaptureSelect™* Fc resin for purification of fragments. One FabRICATOR Z spin column contains sufficient FabRICATOR Z-coupled agarose beads to process 0.5 mg mouse IgG2a and IgG3.
- Purification of homogenous F(ab')2 fragments from mouse IgG2a and IgG3
- Fast - pure fragments in under two hours
- Reliable workflow for preparative applications
Product Composition: FabRICATOR Z Immobilized microspin column containing 0.1 ml settled resin of FabRICATOR Z enzyme immobilized on agarose. CaptureSelect Fc microspn column containing 0.1 ml settled resin of CaptureSelect™ Fc₁ affinity ligand immobilized on agarose. Agarose resins are suspended in 20% ethanol/water (v/v).
The immobilized enzyme and affinity resin helps for hinge digestion of mouse IgG2a and IgG3 and purification of fragments.
One spin column contains sufficient FabRICATOR Z-coupled agarose beads to digest 0.5 mg mouse IgG2a or IgG3.
The FabRICATOR Z Immobilized spin columns and CaptureSelect™* spin columns are supplied in 20% ethanol, shipped cold, and should be stored at 4 to 8 °C upon arrival. Do not freeze the products.
Appearance: White suspension.
Binding capacity: >15 mg hIgG per ml CaptureSelect Fc resin.
Specifications
- Storage Temperature:4...8 °C