ContactUsLinkComponent
VivaGuard® Lancing Device, Germaine Laboratories
VivaGuard® Lancing Device, Germaine Laboratories
Catalog # 76670-428
undefined
VivaGuard® Lancing Device, Germaine Laboratories
Catalog # 76670-428
Supplier Number:  VGD01-381
Specifications
  • Description:
    Lancing device
  • Cat. No.:
    76670-428
Specifications
About this item
Lancing device with five depth settings offers maximum comfort through selecting a proper penetration depth.
  • Easy to carry with a secure-grip body design for safety
  • Smooth gliding mechanism offers reliability and simplicity
  • Convenient storage with five-year shelf life
Unique design and easy to use-featuring a lancet ejector that easily disposes of the used lancet, hands-free, to prevent accidents and for better safety.
Delivery information: One shipping box.