LED Ray Box
LED ray box, 6.25×3.5×3.25''
Catalog # 470344-164
Supplier: Wards
: PH0602ALED
Jump to:
Product Details & Documents
Demonstrate the optical principles of lenses and mirrors.
- Battery operated LED light ray box with seven grooves
- Ideal for classroom demonstrations in physics, optics, reflection, refraction and more
- Includes two slit plates and one lens
- Rugged ABS injection molded enclosure
- Runs substantially cooler than traditional incandescent lamps
This ray box in an ABS injection molded enclosure, and delivers bright light but runs much cooler than conventional incandescent lamp models. It uses the latest, cool ultra bright LED technology giving superior white light. Supplied complete with lens, two slit plates and its own battery power source.
: Batteries are required, but not included.
Specifications
- Description:LED ray box
- L×W×H:6.25×3.5×3.25''