(4-Nitrophenyl)-β-D-glucopyranoside
Catalog # 80110-008
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Size:1 g
- Cat. No.:80110-008
- Supplier no.:487507-1GM
Specifications
About this item
:
:301.25 g/mol
CAS Number:2492-87-7
Formula:C₁₂H₁₅NO₈
MDL Number:MFCD00006593