Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium dodecyl sulfate (SDS) solution 20%
Sodium dodecyl sulfate (SDS) solution 20%
Catalog # 80058-310
Supplier:  MilliporeSigma
CAS Number:  
Sodium dodecyl sulfate (SDS) solution 20%
Catalog # 80058-310
Supplier:  MilliporeSigma
Supplier Number:  428018-200ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic ampoule
  • Size:
    200 ml
  • Cat. No.:
    80058-310
  • Supplier no.:
    428018-200ML

Specifications

About this item

 :
 :288.38 g/mol
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175