IPTG (isopropyl-β-D-thiogalactopyranoside) Hemidioxane Adduct
Catalog # 80056-910
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Size:5 g
- Cat. No.:80056-910
- Supplier no.:420290-5GM
Specifications
About this item
:
:238.31 g/mol
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273