UDP-α-D-N-Acetylglucosamine Disodium Salt
Catalog # 80054-958
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Size:50 mg
- Environmentally Preferable:
- Cat. No.:80054-958
- Supplier no.:670107-50MG
Specifications
About this item
Synonyms: UDP-GlcNAc disodium
:
:651.3174 g/mol
CAS Number:91183-98-1
Formula:C₁₇H₂₅N₃Na₂O₁₇P₂
MDL Number:MFCD00799985