Order Entry
Puerto Rico
ContactUsLinkComponent
eBlot™ L1 Fast Wet Transfer System
eBlot™ L1 Fast Wet Transfer System
Catalog # 76510-444
Supplier:  Genscript
undefined
eBlot™ L1 Fast Wet Transfer System
Catalog # 76510-444
Supplier:  Genscript
Supplier Number:  L00686

Specifications

  • Description:
    eBlot™ L1 fast wet transfer system
  • Size:
    1 unit
  • Cat. No.:
    76510-444

Specifications

About this item

eBlot™ L1 wet protein transfer system is a fast western blotting protein transfer system that combines the high reproducibility and efficiency of the traditional wet protein transfer with the speed and convenience of semi-dry protein transfer. It allows efficient western blot transfer of small, medium, and large molecular weight proteins.

  • Automated fast wet protein transfer: 1 or 2 mini gels in 9 to 17 minutes
  • Full protein size coverage: Outstanding performance for small, medium, and large molecular weight proteins
  • High protein transfer efficiency: With gold-standard wet protein transfer technology
  • Convenience: Unique cassette sandwich setup allows easy and clean operation
  • Flexibility: Customizable program
  • All-inclusive consumable kit: Include precut NC/PVDF membranes and buffers with a long shelf life
  • Highly compatible with different types of precast or homemade mini gels

eBlot™ L1 allows fast and efficient protein transfer of 1 or 2 mini gels at a time. The system comes with conveniently designed transfer stacks/consumables (sold separately). The setup takes less than 2 minutes and can be performed even with the sponge in a dry state (patented).

Why Choose eBlot™ L1? Conventional western blot wet transfers are cumbersome and require experience and a great deal of time spent on consumable preparation. The transfer times usually vary from one hour to overnight. With eBlot™ L1 fast wet protein transfer system, achieve excellent western blot transfer efficiency without sacrificing speed.