ContactUsLinkComponent
Valve Lockout Tagout Kit with Aluminum Safety Padlocks in Toolbox, Brady Worldwide
Valve Lockout Tagout Kit with Aluminum Safety Padlocks in Toolbox, Brady Worldwide
Catalog # 76637-602
Supplier:  Brady Worldwide
undefined
Valve Lockout Tagout Kit with Aluminum Safety Padlocks in Toolbox, Brady Worldwide
Catalog # 76637-602
Supplier:  Brady Worldwide
Supplier Number:  153671
Specifications
  • Color:
    Black
  • Description:
    Valve lockout kit, keyed different
  • Dimensions:
    16.2W×7.3D×7.8"H
  • Material:
    Nylon
  • Cat. No.:
    76637-602
Specifications
About this item
Safely power down faster when you power up your lockout tagout readiness with one convenient, easily accessible and portable kit.
  • OSHA compliance driver
  • Built to last
  • Extremely portable
  • Great for contractors
  • Stores easily in vehicles to travel between jobsites
Designed and assembled exclusively for safety to help users comply with OSHA 29 CFR § 1910.147 control of hazardous energy. All components meet Brady’s highest standards for quality, durability and reliability.
Toolbox keeps padlocks and devices at the ready and goes anywhere. Everything needed to effectively lock out gate, ball, and other valves. Each lock and key are unique, allowing authorized users to have their own individual locks and comply with OSHA’s 'one person, one lock, one key' guidance.
Ordering information: Lockout sit includes: (3) Aluminum safety lockout padlocks, (1) lockout toolbox, (2) 1" lockout hasps, (1) cable lockout with 8' metal cable, (1) universal valve lockout with blocking arm, (1) gate valve lockout 1 to 2.5", (1) gate valve lockout 2.5 to 5", (1) gate valve lockout 5 to 6.5", (1) ball valve lockout, (10) lockout tags, and (10) zip ties.