Electrical Lockout Station with Safety Lockout Padlocks, Brady Worldwide
Supplier: Brady Worldwide
Certificates
About this item
Safely power down faster when powering up lockout tagout readiness with this fully outfitted, easily installed, organized and accessible electrical lockout station.
- OSHA compliance driver
- Built to last
- Ultimate accessibility
Designed and assembled exclusively for safety to help users comply with OSHA 29 CFR § 1910.147 control of hazardous energy. All components meet Brady’s highest standards for quality, durability and reliability. Place locks, tags and devices at the point of need to help authorized workers maintain safety and compliance. Comes with predrilled holes for easy wall mounting, and a carrying handle when it is needed on the go.
Ordering information: Lockout station 153686 includes: (6) nylon safety lockout padlocks, keyed alike, (1) lock and device station, (2) 1" lockout hasps, (2) 1.5" lockout hasps, (1) cable lockout with 8' metal cable, (2) clamp-on circuit breaker lockout 120/277 V, (1) clamp-on circuit breaker lockout 480/600 V, (2) universal multipole breaker lockouts, (1) 3-in-1 plug lockout, (2) wall switch lockouts, (20) lockout tags, and (20) zip ties.
Lockout station 153685 includes: (6) nylon safety lockout padlocks, keyed different, (1) lock and device station, (2) 1" lockout hasps, (2) 1.5" lockout hasps, (1) cable lockout with 8' metal cable, (2) clamp-on circuit breaker lockouts 120/277 V, (1) clamp-on circuit breaker lockout 480/600 V, (2) universal multipole breaker lockouts, (1) 3-in-1 plug lockout, (2) wall switch lockouts, (20) lockout tags, and (20) zip ties.
Lockout station 153685 includes: (6) nylon safety lockout padlocks, keyed different, (1) lock and device station, (2) 1" lockout hasps, (2) 1.5" lockout hasps, (1) cable lockout with 8' metal cable, (2) clamp-on circuit breaker lockouts 120/277 V, (1) clamp-on circuit breaker lockout 480/600 V, (2) universal multipole breaker lockouts, (1) 3-in-1 plug lockout, (2) wall switch lockouts, (20) lockout tags, and (20) zip ties.