Electrical and Valve Lockout Tagout Kit with Nylon Safety Lockout Padlocks in Duffel Bag, Brady Worldwide
Catalog # 76637-598
Supplier: Brady Worldwide
Specifications
- Color:Blue
- Description:Valve Lockout Kit, keyed different
- Dimensions:10H×16.5W×8.5"D
- Material:Polyester
- Cat. No.:76637-598
Specifications
About this item
Safely power down faster when powering up lockout tagout readiness with this convenient, easily accessible and portable lockout kit.
- OSHA compliance driver
- Built to last
- Extremely portable
- Great for contractors
- Sores easily in vehicles to travel between jobsites
Designed and assembled exclusively for safety to help users comply with OSHA 29 CFR § 1910.147 Control of Hazardous Energy. All components meet Brady’s highest standards for quality, durability and reliability.
Duffel keeps padlocks and devices at the ready and goes virtually anywhere. Organize, transport, and deploy everything needed to lock out breakers, plugs, and valves in a multitude of types and sizes with this robust kit. Each lock and key are unique, allowing authorized users to have their own individual locks and comply with OSHA’s 'one person, one lock, one key' guidance.
Ordering information: Lockout Kit includes: (3) Nylon Safety Lockout Padlocks, (1) Lockout Duffel, (2) Lockout Pouches, (2) Lockout Hasps, (1) Cable Lockout with 8' Metal Cable, (1)Universal Valve Lockout with Blocking Arm, (1) Gate Valve Lockout 1 - 2.5", (1) Gate Valve Lockout 2.5- 5", (1) Gate Valve Lockout 5 - 6.5", (1) Ball Valve Lockout, (2) Clamp-on Circuit Breaker Lockout 120/277 V, (1) Clamp-on Circuit Breaker Lockout 480/600V, (2) Universal Multipole Breaker Lockout, (1) 3-in-1 Plug Lockout, (1) Wall Switch Lockout, (10) Lockout Tags, and (10) Zip Ties.