About this item
Eppendorf ep Dualfilter T.I.P.S.® 384 Racks deliver cutting-edge filter pipette tips in 'PCR clean and Sterile' grade, specifically designed for 384-well plate applications, ensuring utmost precision and contamination protection in high-throughput labs. ep Dualfilter T.I.P.S.® 384 pipette tips build a system together with the Research® plus and Xplorer® plus 16- and 24-channel pipettes as well as 20 and 100 µl Move It® adjustable tip spacing pipettes for utmost safety and reliability.
- Innovative SOFTattach technology allows for reliable tip fit and tightness while operating forces are reduced to a minimum
- Fill a whole 384-well plate without loosening pipette tips
- Tip shape is adjusted to perfectly fit the wells of 384-well plates
- Dual protection for pipette and sample against aerosols and biomolecules
- Available as racks and in purity grades PCR clean and sterile
- Modern slim rack design with reclosable lid
- Improved sustainability through 20% lower polypropylene consumption per rack
- ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
Perfect tip alignment is guaranteed by extraordinary coaxiality for safe plate maneuvering and liquid transfer.
ep Dualfilter T.I.P.S.® 384 with innovative SOFTattach technology are the first filter pipette tips with elastic forming grooves that allow the tips to stretch as much as necessary to enable a perfect seal, while also keeping operating forces to a minimum. These tips were designed to be used with Eppendorf 16- and 24-channel pipettes as well as 20 and 100 µl Move It® adjustable tip spacing pipettes in any application involving 384-well plates. Use this perfectly optimized pipette and filter tip system to benefit from outstanding tip fit and tightness combined with reduced tip attachment and ejection forces. Offering an extremely fine tip shape and the ultimate coaxiality for a perfect tip alignment.
ep Dualfilter T.I.P.S.® 384 pipette tips for 384-well plates are the latest addition to the premium ep Dualfilter T.I.P.S.® family from Eppendorf. Both provide the ultimate protection of pipette and sample against unintentional and unnoticed contamination by aerosols, drops, and splashes. The aerosol barrier pipette tips are optimized for Eppendorf pipettes and meet EN ISO 8655 requirements. Optimal wetting properties, high transparency and special, certified purity grades are the visible expression of Eppendorf product and production philosophy.
Specifications
- Catalog No:
- 76311-152
- 76311-150
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001