Specifications
- Description:VacuTip II
- Environmentally Preferable:
- Cat. No.:MSPP-519500044
Specifications
About this item
Glass microcapillary for holding suspension cells (eg. oocytes or blastocysts).
- 60 μm inner diameter, 110 µm outer diameter
- 25° tip angle
- Flange length 0.5 mm
- Smooth rounded tip end for gentle holding
- Sterilised by gamma irradiation
- Non cytotoxic (as proven by mouse embryo assay)
- Capillary safe for optimum protection
Caution: For research use only.