Automated dehumidification system offers convenience.
- Set relative humidity range (RH) between 25 and 55%
- Built-in digital hygrometer allows easy monitoring of RH
- View samples through acrylic viewing panels
Keeping a constant RH level has never been easier. The electronic dehumidification system includes a permanent desiccant and a hygrometer to keep the relative humidity level between 25 and 55%. The desiccant automatically recycles by discharging absorbed moisture through an external shutter. Once it is recycled, desiccant is ready to absorb moisture again. Safely keep humidity sensitive products such as optical equipment, electronic parts, PCB boards, and reagents.
Aluminum frame and stainless steel shelves offer excellent durability. Stainless steel shelves have holes to ensure uniform humidity throughout chamber. A lock and key allow you to securely lock the cabinet to prevent unauthorized access.
- Catalog No:
- 76640-524
- 76640-528
- Electrical:
- 110 VAC, 50/60 Hz
- 110 VAC, 50/60 Hz
- Ext. Dimensions:
- 43.3×40×43.3 cm (17×15.7×17")
- 50.2×38×58.1 cm (19.8×15×22.9")
- Int. Dimensions:
- 33×33×33.5 cm (13×13×13.2")
- 47.8×31×40.5 cm (18.8×12.2×15.9")
- Int. Volume:
- 50 L
- 70 L