Tissue Culture Plates, Ultra Optically Clear, Azenta Life Sciences
Supplier: AZENTA US, INC
Certificates
About this item
The main advantage of using ultra optically clear plates is in the optically ultra-clear base of the plate, which gives superior results by delivering low absorbance and high transmission, together with low background signals.
- Free from DNase, RNase and human genomic DNA
- Ultra-clear base improves transmission for low wavelengths
- Allows DNA measurements at 260/280 nm wavelengths in a medium or high throughput
- Peel-back film on the base for scratch free surface
- Optimum signal-to-noise ratios
- Alphanumeric grid reference to aid well and sample identification
- Suitable for adhesive and heat sealing
Due to its incredible optical clarity, the ultraclear base of the ultra optically clear plate delivers improved transmission of signals at low wavelengths compared to standard optical films.