Heat Sealing Foils, Peelable
Supplier: AZENTA US, INC
Certificates
About this item
The peel heat seal is a laminate seal compatible with polypropylene plates, suitable for low temperature storage, high temperature uses and PCR.
- Peelable from polypropylene and COC plates
- Good solvent resistance including DMSO
- Autoclavable (121 °C)
- Suitable for short term room temperature storage
- Suitable for very low temperature storage
- Temperature range: –80 to +90 °C
- Non sterile
Heat sealing offers a 100% effective method for plate sealing for a complete seal integrity, as well as being quick and cost effective. Peelable foil heat seal forms a complete seal to a plate enabling very low temperature uses, including very low temperature storage, and high temperature uses, such as PCR (when used with a pressurized heated lid).
The sheet seals fit all standard SBS footprint PCR and qPCR plates, microplates, assay and storage plates, from 12 to 1536 wells.
Ordering information: Available as sheets for use with manual and semiautomated sealers. Available in multiple roll formats compatible with specified automated heat sealers. Gamma treatment available upon request.