Order Entry
Puerto Rico
ContactUsLinkComponent
2D Coded Sample Storage Tubes, 0.8 ml, 96-format, External Thread, Azenta Life Sciences
2D Coded Sample Storage Tubes, 0.8 ml, 96-format, External Thread, Azenta Life Sciences
Supplier:  AZENTA US, INC
Certificates
undefined
2D Coded Sample Storage Tubes, 0.8 ml, 96-format, External Thread, Azenta Life Sciences
Supplier:  AZENTA US, INC
Certificates

Specifications

  • Catalog No:
  • 76636-764
  • 76637-038
  • 76636-560
  • 76636-722
  • Height with cap:
  • 40.3 mm
  • 40.3 mm
  • 40.3 mm
  • 40.3 mm
  • Ext.Ø with cap:
  • 8.7 mm
  • 8.7 mm
  • 8.7 mm
  • 8.7 mm
  • Centre to centre:
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • Height in rack:
  • 41.2 mm
  • 41.2 mm
  • 41.2 mm
  • 41.2 mm
  • Overall rack height incl. lid:
  • 43.9 mm
  • 43.9 mm
  • 43.9 mm
  • 43.9 mm

Specifications

Empty racks

Product Information