Storage Microplate, 96 Square Well, KingFisher Style, Azenta Life Sciences
Catalog # 76637-994
Supplier: AZENTA US, INC
Specifications
- Well volume:200 µl
- Color:Clear
- Description:96-well storage microplate KingFisher style
- Material:PP
- Cat. No.:76637-994
- Well bottom shape:V-bottom
- Bottom style:V-shape
Specifications
About this item
Suitable for use with KingFisher Flex, Apex and Presto.
- Made from polypropylene, a material with very low biomolecular binding characteristics, a high temperature tolerance and resistance to many standard laboratory chemicals, including DMSO
- Alphanumeric grid reference to aid well and sample identification
- SBS footprint
- Compatible with automation
- Square wells, to make the best use of space, and to improve mixing
- Stackable
Compatible with KingFisher system.