Order Entry
Puerto Rico
ContactUsLinkComponent
 
Storage Microplate, 384 Square Well, Deep Well
Catalog # 76638-196
Supplier:  AZENTA US, INC
undefined
Storage Microplate, 384 Square Well, Deep Well
Catalog # 76638-196
Supplier:  AZENTA US, INC
Supplier Number:  4TI-0147

Specifications

  • Well volume:
    190 µl
  • Color:
    Clear
  • Description:
    384 deep well storage microplate
  • Material:
    PP
  • Cat. No.:
    76638-196
  • Well bottom shape:
    V-bottom
  • Bottom style:
    V-shape

Specifications

About this item

These storage plates are designed for high density sample collection and storage for a wide array of applications within cell biology, molecular biology and drug discovery. Clear plate, perfect for storage applications.

  • Made from polypropylene, a material with very low biomolecular binding characteristics, a high temperature tolerance and resistance to many standard laboratory chemicals, including DMSO
  • Autoclavable and stackable
  • Free from RNase, DNase, human genomic DNA and endotoxins
  • Alphanumeric grid reference to aid well and sample identification
  • SBS footprint
  • Square wells, to make the best use of space, and to improve mixing
  • Temperature range for use: –80 to 120 °C
  • Suitable for adhesive and heat sealing, and as a collection plate from filter systems

V-shaped bottom is best for precipitation, centrifugation and small volume recovery due to the V-shaped bottom, as the liquid gathers at the lowest point of the V for easy pipetting.

Accessories information: 384 square well sealing cap mat, clear silicone, for use with square 384 well microplates and deep well storage microplates.

Sealing Mat

Product Information