Sealing Cap Strips, Flat, Crystal Clear, Azenta Life Sciences
Supplier: AZENTA US, INC
Certificates
About this item
These strips of 8 flat crystal clear optical caps are designed for low volume applications such as low volume qPCR.
- Very tight sealing
- Made of a special polymer with improved optical properties leading to high transmission rates
- Ideally suited for small samples with low signal intensity
- Reduced shrinking during heating and cooling phases
Due to their improved optical properties and evaporation-safe fit, the strips are ideally suited for applications where small volumes are used, e.g., low volume qPCR.