Sealing Cap Strips, Azenta Life Sciences
Supplier: AZENTA US, INC
Certificates
About this item
These clear polypropylene sealing caps, available as strips of 8 caps (domed or flat optical) and strips of 12 caps (flat optical) are molded from virgin polypropylene and are compatible with our tube strips and 96 well PCR plates.
- Easy to apply
- Large end tabs for easy removal
- Labeled for orientation
Flat caps are optically clear for fluorescence detection (e.g. qPCR).