Order Entry
United States
ContactUsLinkComponent
Choice® SatPax 500 Pre-wetted 60% Cellulose/40% Polyester Nonwoven Cleanroom Wipers, Berkshire
Choice® SatPax 500 Pre-wetted 60% Cellulose/40% Polyester Nonwoven Cleanroom Wipers, Berkshire
Catalog # 76638-266
Supplier:  Berkshire
undefined
Choice® SatPax 500 Pre-wetted 60% Cellulose/40% Polyester Nonwoven Cleanroom Wipers, Berkshire
Catalog # 76638-266
Supplier:  Berkshire
Supplier Number:  SPXCHN50018

Specifications

  • Description:
    Pre-wetted, nonwoven cleanroom wiper
  • Environmentally Preferable:
  • Cat. No.:
    76638-266
  • Packaging:
    180 sheets/pack (C-fold)
  • W×L:
    15.2×20.3 cm (6×8")
  • Solvent:
    6% IPA, 94% DI water, 45% saturation

Specifications

About this item

Choice® SatPax 500 wipes are composed of a hydroentangled nonwoven blend of 60% cellulose and 40% polyester. This pre-wetted format provides a cost-effective and easy-to-use solution versus traditional bulk handling of solvents.

  • Excellent combination of synthetic polyester strength and cleanliness, with the absorbent characteristics of cellulose
  • Smooth and durable with good wet strength
  • Low extractable levels and particle counts
  • Chemically compatible with common cleaning and disinfecting solutions
  • Reduces VOC emissions

Choice® value tiered category of wipers offers all the performance consistency of higher priced wipers. Choice® 500 wipes are composed of a hydroentangled nonwoven blend of 60% cellulose and 40% polyester, with three saturation levels of isopropyl alcohol and DI water. This pre-wetted format provides a cost-effective and easy-to-use solution versus traditional bulk handling of solvents, maintenance of squirt bottles, and inconsistent wetting and cleaning associated with prepping dry wipers.

Certifications: ISO Class 5 – 8. Cleanroom Class 100 – 100,000. EU Grade B/C/D.