Semi-Automated Sheet Heat Sealer (formerly IntelliXseal™ SA), Azenta Life Sciences
Catalog # 76638-864
Supplier: AZENTA US, INC
Specifications
- Power:Supply: 100 - 240 V AC +/- 10% 50/60 Hz; consumption: 300 W (max.)
- Description:Semi-automated individual access heat sealer
- Dimensions:181×275×310 mm
- Weight:9 kg
- Sealing Time:1 - 10 sec
- Sealing temperature range:60 - 200 °C
- Cat. No.:76638-864
Specifications
About this item
Semi-automated sheet heat sealer compatible with a wide range of seals and microplates, PCR and assay plates, as well as storage plates of differing designs and heights.
Enables optimization of any seal/plate type and is compatible with all SBS microplates
Easy to use with a simple, three button operation
- Real-time temperature display with variable time and temperature controls
- Auto stand-by function and switch-off mode to help conserve energy
- RS-232 port enables unit integration within robotic systems
- Small footprint takes minimal bench space
Connection: RS-232 serial port
Certifications:
CE marked.
Ordering information:
12-month warranty.
Packaging:
One sealer system per case.