Anti-Caspase 1 Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
Certificates
About this item
Caspase 1 and is expressed as a proenzyme in many tissues. Active Caspase 1 is a tetramer of two subunits p10 and p20 in 2:2 ratio. Overexpression of caspase 1 can induce apoptosis in fibroblast, which can be inhibited by overexpression of Crm A, a protein from pox virus.
Specifications
- Catalog No:
- 76635-194
- 76635-196
- 76635-192
- Antigen Name:
- Caspase 1
- Caspase 1
- Caspase 1
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Rabbit