Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Factro XIIIa Mouse Monoclonal Antibody [clone: AC-1A1]
Anti-Factro XIIIa Mouse Monoclonal Antibody [clone: AC-1A1]
Catalog # 76634-394
Anti-Factro XIIIa Mouse Monoclonal Antibody [clone: AC-1A1]
Catalog # 76634-394
Supplier Number:  PDM141

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Factro XIIIa
  • Clonality:
    Monoclonal
  • Clone:
    AC-1A1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Size:
    6 ml
  • Cat. No.:
    76634-394

Specifications

About this item

This antibody is specific to 160 kDa protein known as Factor XIII-A. Factor XIII is a β-globulin found in plasma and is composed of two subunits, Factor XIII-A and Factor XIII-B. Factor XIII-A is the catalytic subunit and is a dimer of M.W. 160 kDa. Factor XIII is a dermal dendrocyte marker and v variable reaction with these types of tumors.