Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Synaptophysin Rabbit Monoclonal Antibody [clone: SP11]
Anti-Synaptophysin Rabbit Monoclonal Antibody [clone: SP11]
Certificates
Anti-Synaptophysin Rabbit Monoclonal Antibody [clone: SP11]

Specifications

  • Catalog No:
  • 76634-762
  • 76634-764
  • 76634-766
  • Antigen Name:
  • Synaptophysin
  • Synaptophysin
  • Synaptophysin
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • SP11
  • SP11
  • SP11
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG

Specifications