Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MUM1 protein Mouse Monoclonal Antibody [clone: MUM1p]
Anti-MUM1 protein Mouse Monoclonal Antibody [clone: MUM1p]
Catalog # 76634-508
Anti-MUM1 protein Mouse Monoclonal Antibody [clone: MUM1p]
Catalog # 76634-508
Supplier Number:  PDM420

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    MUM1 protein
  • Clonality:
    Monoclonal
  • Clone:
    MUM1p
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    6 ml
  • Cat. No.:
    76634-508

Specifications

About this item

Multiple myeloma oncogene-1 (MUM1) is a 50 kDa protein encoded by the MUM1 gene. This antibody stains MUM1 protein, which is expressed in a subset of B cells in the light zone of the germinal center, plasma cells, activated T cells and a wide spectrum of related hematolymphoid neoplasms. This antibody is useful in the sub classification of lymphoid malignancies.