Anti-HHV8 Rat Monoclonal Antibody [clone: LN53]
Catalog # 76634-506
Supplier: Diagnostic Biosystems
Specifications
- Antibody Type:Primary
- Antigen Name:HHV8
- Clonality:Monoclonal
- Clone:LN53
- Conjugation:Unconjugated
- Host:Rat
- Isotype:IgG2c
- Reactivity:Human
- Size:6 ml
- Cat. No.:76634-506
Specifications
About this item
This antibody stains with ORF-73 of human herpesvirus 8 (HHV8). HHV8 is the likely etiological agent of Kaposi sarcoma (KS). HHV8 encodes a latent nuclear antigen (LNA), which is the product of the viral gene ORF 73. LNA is capable of forming a complex with retinoblastoma susceptibility gene product, which may be related to its oncogenic activity.