Anti-SOX10 Mouse Antibody [clone: 20B7]
Catalog # 76634-542
Supplier: Diagnostic Biosystems
Specifications
- Antibody Type:Primary
- Antigen Name:SOX10
- Clone:20B7
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:6 ml
- Cat. No.:76634-542
Specifications
About this item
SOX10 is widely expressed in normal human tissues including melanocytes and breast tissue. SOX10 is also an important marker in malignant tumors such as melanoma, breast carcinoma, gliomas, and benign tumors such as schwannomas.SOX-10 is considered as a very reliable marker for recognizing residual desmoplastic melanomas. In normal tissues, it is expressed in Schwann cells, melanocytes and myoepithelial cells of salivary, bronchial and mammary glands. SOX-10 expression is also observed in mast cells.