Anti-Smooth muscle myosin Mouse Monoclonal Antibody [clone: SMMS-1]
76634-450
- Antibody Type:Primary
- Antigen Name:Smooth muscle myosin
- Clonality:Monoclonal
- Clone:SMMS-1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:6 ml
- Cat. No.:76634-450
Monoclonal anti-smooth muscle myosin reacts with myosin heavy chain polypeptides of 200 and 204 kDa. It does not react with skeletal, cardiac, or non-muscle myosin. In immunohistochemistry it stains vascular and visceral smooth muscle cells and cells that have smooth muscle-like characteristics.