To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Actin sarcomeric
- Clonality:Monoclonal
- Clone:5C5
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Size:6 ml
- Cat. No.:76634-348
This antibody is specific to α-smooth muscle isoform of actin. It reacts with smooth muscle cells of vessels and different parenchymes. This antibody does not cross-react with β and γ-cytoplasmic, α-sarcomeric and α-myocardial actin isoforms.