Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-DOG1.1 Mouse Monoclonal Antibody [clone: DOG1.1]
Anti-DOG1.1 Mouse Monoclonal Antibody [clone: DOG1.1]
Certificates
Anti-DOG1.1 Mouse Monoclonal Antibody [clone: DOG1.1]

Specifications

  • Catalog No:
  • 76633-974
  • 76633-970
  • 76633-972
  • Antigen Name:
  • Dog1.1
  • Dog1.1
  • Dog1.1
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • DOG1.1
  • DOG1.1
  • DOG1.1
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG
  • IgG
  • IgG

Specifications