Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-IgM Rabbit Polyclonal Antibody
Anti-IgM Rabbit Polyclonal Antibody
Certificates
Anti-IgM Rabbit Polyclonal Antibody

Specifications

  • Catalog No:
  • 76634-982
  • 76634-984
  • Antigen Name:
  • IgM
  • IgM
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit

Specifications