About this item
This antibody is specific to 45-60 kDa proteins identified as tau proteins. This antibody does not cross react with tubulin or other microtubule associated proteins. This antibody recognizes a phosphorylation-independent epitope in amino acids 404 to 441 (human). Tau antibody stains human neurofibrillary tangles, neutrophil threads and neurotic plaques associated with Alzheimer’s disease.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76633-998
- 76633-996
- 76633-994
- Antigen Name:
- Tau (neurofibrillary tangles marker)
- Tau (Neurofibrillary Tangles Marker)
- Tau (Neurofibrillary Tangles Marker)
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- Tau46
- Tau46
- Tau46
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1