Bacto™ Beef Extract, Desiccated, Gibco™ (a part of Thermo Fisher Scientific)
76628-786
:
- Description:Bacto™ Beef extract, desiccated
- Size:500 g
- Cat. No.:76628-786
- Source:Bovine
Bacto Beef extract, desiccated, is an animal origin (AO) peptone derived from an infusion of beef. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins.
- Derived from infusion of beef
- Provides a rich source of nutrientsDesiccated
- Dried form of Beef Extract paste
- Applications-Intended to replace aqueous infusion of meat in microbiological culture media
Dried from beef extract paste, Bacto Beef Extract, desiccated, has a granular appearance and different nutritional profile than other beef extract products in our portfolio. Customers have used Bacto Beef extract, desiccated, in a variety of microbial fermentation cell lines including Clostridium species, Escherichia coli, Bacillus subtilis, Pasteurella multocida, and Streptococcus species to produce numerous animal health vaccines.