ContactUsLinkComponent
Difco™ PPLO Broth, Gibco™ (a part of Thermo Fisher Scientific)
Difco™ PPLO Broth, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # 76628-918
CAS Number:  
Difco™ PPLO Broth, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # 76628-918
Supplier Number:  255410
CAS Number:  
Specifications
  • Description:
    Difco™ PPLO broth, animal origin (Porcine)
  • Size:
    10 kg
  • Cat. No.:
    76628-918
Specifications
About this item
Difco PPLO Broth is used for isolating and cultivating Mycoplasma, when supplemented with nutritive enrichments.
  • Mixture of meat digests, peptones, beef extract, and yeast extract that provide the nitrogen, vitamins, amino acids, and carbon for optimal growth of Mycoplasma species
  • When supplemented with nutritive enrichments, are used for isolating and cultivating Mycoplasma
  • Used in animal health applications for the propogation of Mycoplasma species such as, Mycoplasma mycoides, Mycoplasma gallisepticum, and others for vaccine production