PCR Tube Strips, 4-Well, Rotor-Gene® Style with Caps, Azenta Life Sciences
76638-170
- Volume:0.1 ml
- Color:Clear
- Description:4-well PCR tube strips, Rotor-Gene® style, with strips of caps
- Cat. No.:76638-170
Designed for Qiagen/Corbett Rotor-Gene® instruments.
- Free from DNase, RNase, human genomic DNA,and endotoxin/pyrogen
- Optically clear tubes ideally suited for qPCR
- Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
- Low profile: <0.1 ml (100 µl) working volume, 0.2 ml (200 µl) total well capacity
- Frosted cap extensions allow for easy handling and labeling
- Tube and cap strips can be separated for individual use
- Pack of 250 strips of tubes and caps sufficient for 1,000 reactions
Azenta offers a range of PCR consumables for low to medium throughput applications. These PCR tubes and caps are molded from virgin polypropylene to prevent any PCR background signal.