ContactUsLinkComponent
ImmunoHisto-Sealer™ Preservation solution, Diagnostic BioSystems
ImmunoHisto-Sealer™ Preservation solution, Diagnostic BioSystems
Catalog # 76633-068
CAS Number:  
ImmunoHisto-Sealer™ Preservation solution, Diagnostic BioSystems
Catalog # 76633-068
Supplier Number:  K076
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    30 ml
  • Cat. No.:
    76633-068
Specifications
About this item
ImmunoHisto-Sealer™ is a preservation solution that acts as a sealant to improve tissue morphology and stain over an extended period of time. When applied to the stained section, specimens can be permanently mounted without chromogens fading. Additional benefits of the ImmunoHisto-Sealer™ include: saves hands on time when mounting slides, avoids the use of toxic chemicals such as xylene and eliminates accumulation of hazardous waste. This product is designed for tissue specimens stained with peroxidase, alkaline phosphatase-based systems and various chromogens. After treating with ImmunoHisto-Sealer, water solution chromogens can be mounted in Permanent Mounting Media