- Description:StatFreeze™ Freezespray, 152a refrigerant
- Size:1 Case (6 per case)
- Environmentally Preferable:
- Cat. No.:76548-222
- Volume:9 oz
Ozone safe, does not contain CFC’s OR HCFC’s. Used to quickly cool paraffin blocks and fresh tissue in frozen sections. Sleek bottle is easy to hold and equipped with trigger nozzle and 5” tube to help direct spray.