PolarStat™ Embedding Medium, StatLab
Product Details & Documents
An aqueous-based frozen section embedding medium designed to support tissue blocks in cryostat sectioning. Formulated to promote rapid freezing, enhanced sectioning and consistent results at a working temperature of -20C. 4 oz bottle.
Packaging: Manufactured in the USA
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:PolarStat™ Embedding Medium
- Size:4 oz