X63-saporin is an antibody conjugate comprising of the non-specific monoclonal IgG1 antibody X63, chemically conjugated via a reducible disulfide bridge to the ribosome-inactivating protein saporin, purified from saponaria officinalis. Antibody clone X63 has no known binding ability, and thus can be used as negative control antibody. In combination with saporin, X63-saporin is a useful negative control for targeting IgG1-saporin conjugates such as MC192-saporin.
- Non-reactive (negative control)
X63-saporin is an antibody conjugate comprising of the non-specific monoclonal IgG1 antibody X63, chemically conjugated via a reducible disulfide bridge to the ribosome-inactivating protein saporin, purified from saponaria officinalis. Antibody clone X63 has no known binding ability, and thus can be used as negative control antibody. In combination with saporin, X63-saporin is a useful negative control for targeting IgG1-saporin conjugates such as MC192-saporin.
Other applications: Antibody clone X63 has no known binding ability, and thus can be used as negative control antibody.
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: X63
Conjugation: Saporin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
- Catalog No:
- 76545-478
- 76545-476
- 76545-474
- Antigen Symbol:
- SAP6
- SAP6
- SAP6
- Antigen Name:
- Non-specific Control IgG, Mouse Monoclonal Antibody (Saporin)
- Non-specific Control IgG, Mouse Monoclonal Antibody (Saporin)
- Non-specific Control IgG, Mouse Monoclonal Antibody (Saporin)
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Saporin
- Saporin
- Saporin
- Clone:
- X63
- X63
- X63
- Reactivity:
- Species independent
- Species independent
- Species independent
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- UniprotID #P20656
- UniprotID #P20656
- UniprotID #P20656
- Isotype:
- IgG1
- IgG1
- IgG1
- Amino Acid Number:
- 299 amino acids
- 299 amino acids
- 299 amino acids
- Form:
- Lyophillized
- Lyophillized
- Lyophillized
- Molecular Weight:
- 30 kDa
- 30 kDa
- 30 kDa
- Purification:
- Conjugate was purified by ion-exchange chromatography. Purity >90% by non-reducing SDS-PAGE
- Conjugate was purified by ion-exchange chromatography. Purity >90% by non-reducing SDS-PAGE
- Conjugate was purified by ion-exchange chromatography. Purity >90% by non-reducing SDS-PAGE
- Storage Buffer:
- Reconstitute in sterile-filtered, ultrapure water
- Reconstitute in sterile-filtered, ultrapure water
- Reconstitute in sterile-filtered, ultrapure water
- Storage Temperature:
- 2…8 °C (lyophilized); <–70 °C (liquid)
- 2…8 °C (lyophilized); <–70 °C (liquid)
- 2…8 °C (lyophilized); <–70 °C (liquid)
- Shipping Temperature:
- 25 °C
- 25 °C
- 25 °C