FrameStar® PCR Plates, 96-Well, Semi-Skirted, ABI® Style, Azenta Life Sciences
Supplier: AZENTA US, INC
Certificates
About this item
High profile, 0.2 ml polypropylene wells, polycarbonate frame, cut corner A12, working volume: <200 µl, total well capacity: 300 µl. Ideal for use with robotic systems. Ideal for ABI® thermal cyclers and sequencers. Compatible with standard multichannel pipettes.
- Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
- Stackable
- Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
- Rigid polycarbonate frame for added mechanical stability
- Significantly reduced thermal expansion and sample evaporation for improved consistency in PCR results
- Improved seal integrity due to thermal stability of frame
- Alphanumeric grid reference to aid well and sample identification
- Recommended for low volume PCR
Specifically designed to be directly compatible with all major thermal cyclers, this plate can be used directly in ABI® 96 well instruments without the need for any adapters.
The rigid FrameStar 2-component design eliminates warping and distortion during PCR, making it ideal for use with robotic systems. The semi-skirt allows for labeling or barcoding for sample tracking.
Ordering information: Ultra-low DNA binding option available. Available barcoded upon request.
Packaging: 50 plates per case.
Related Information
FrameStar® PCR Plates, 96-Well, Semi-Skirted, ABI® Style, Azenta Life Sciences_01 FrameStar® PCR Plates, 96-Well, Semi-Skirted, ABI® Style, Azenta Life Sciences_02 FrameStar® PCR Plates, 96-Well, Semi-Skirted, ABI® Style, Azenta Life Sciences_03 FrameStar® PCR Plates, 96-Well, Semi-Skirted, ABI® Style, Azenta Life Sciences_04