Orders LinkContactUsLinkComponent
Sol-M® Oral Syringes (O-Ring Type)
Sol-M® Oral Syringes (O-Ring Type)
Sol-M® Oral Syringes (O-Ring Type)
Certificates
Sol-M® Oral Syringes (O-Ring Type)
Sol-M® Oral Syringes (O-Ring Type)
8 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Sol-M Oral Syringe has been specifically designed for use with oral medications.
  • Clear graduation markings for accurate medication delivery
  • Not made with natural rubber latex
  • Optional amber barrel designed for light-sensitive drugs, in compliance with United States Pharmacopeia (USP) requirements
Special tip design will not accept Luer connectors, preventing medication errors and the accidental intravenous injections of oral liquids.
Documents
Product Family Options
Product Information
  • Volume
    Availability
    Price
  • 76582-684
    Syringe with tip cap, amber
    Syringe with tip cap, amber
    1 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $38.28
    $38.28
    Log in for availability
    Case of 400
    Case of 400 - $153.12
    $153.12
    Box of 100
       $38.28
    Case of 400
       $153.12
    Supplier Number: 21001OA
  • 76582-686
    Syringe with tip cap, clear
    Syringe with tip cap, clear
    1 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $33.98
    $33.98
    Log in for availability
    Case of 400
    Case of 400 - $135.92
    $135.92
    Box of 100
       $33.98
    Case of 400
       $135.92
    Supplier Number: 21001OC
  • 76582-692
    Syringe with tip cap, amber
    Syringe with tip cap, amber
    3 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 400
    Box of 400 - $38.02
    $38.02
    Log in for availability
    Case of 1600
    Case of 1600 - $152.08
    $152.08
    Box of 400
       $38.02
    Case of 1600
       $152.08
    Supplier Number: 21003OA
  • 76582-694
    Syringe with tip cap, clear
    Syringe with tip cap, clear
    3 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 400
    Box of 400 - $43.17
    $43.17
    Log in for availability
    Case of 1600
    Case of 1600 - $172.68
    $172.68
    Box of 400
       $43.17
    Case of 1600
       $172.68
    Supplier Number: 21003OC
  • 76582-700
    Syringe with tip cap, amber
    Syringe with tip cap, amber
    5 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 400
    Box of 400 - $51.52
    $51.52
    Log in for availability
    Case of 1600
    Case of 1600 - $206.08
    $206.08
    Box of 400
       $51.52
    Case of 1600
       $206.08
    Supplier Number: 21005OA
  • 76582-702
    Syringe with tip cap, clear
    Syringe with tip cap, clear
    5 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 400
    Box of 400 - $50.33
    $50.33
    Log in for availability
    Case of 1600
    Case of 1600 - $201.32
    $201.32
    Box of 400
       $50.33
    Case of 1600
       $201.32
    Supplier Number: 21005OC
  • 76582-676
    Syringe with tip cap, amber
    Syringe with tip cap, amber
    10 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $88.89
    $88.89
    Log in for availability
    Case of 400
    Case of 400 - $355.56
    $355.56
    Box of 100
       $88.89
    Case of 400
       $355.56
    Supplier Number: 20010OA
  • 76582-708
    Syringe with tip cap, clear
    Syringe with tip cap, clear
    10 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 100
    Box of 100 - $88.81
    $88.81
    Log in for availability
    Case of 400
    Case of 400 - $355.24
    $355.24
    Box of 100
       $88.81
    Case of 400
       $355.24
    Supplier Number: 21010OC