Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Biological Buffers
GenScript® PAGE-MASTER Protein Standard (for SDS-PAGE), GenScript
GenScript® PAGE-MASTER Protein Standard (for SDS-PAGE), GenScript
GenScript® PAGE-MASTER Protein Standard (for SDS-PAGE), GenScript
Catalog # 76197-946
Supplier:
Genscript
CAS Number:
Expand image
GenScript® PAGE-MASTER Protein Standard (for SDS-PAGE), GenScript
Catalog # 76197-946
Supplier:
Genscript
Supplier Number:
M00516
CAS Number:
Specifications
Description:
PAGE-MASTER protein standard (for SDS-PAGE)
Cat. No.:
76197-946
Specifications
About this item
Product Details
Product Details