Order Entry
Puerto Rico
ContactUsLinkComponent
EnzyFluo™ Human/Mouse LKB1(S431) Phosphorylation ELISA Kit, BioAssay Systems
EnzyFluo™ Human/Mouse LKB1(S431) Phosphorylation ELISA Kit, BioAssay Systems
Catalog # 76545-520
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyFluo™ Human/Mouse LKB1(S431) Phosphorylation ELISA Kit, BioAssay Systems
Catalog # 76545-520
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  ELKB1-100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 tests
  • Cat. No.:
    76545-520

Specifications

About this item

Determination of LKB1(S431) phosphorylation status in whole cells and evaluation of pathway modulation by activators, inhibitors, siRNA etc.

  • New and improved: Total assay time reduced from the standard 9 hours to 5 hours
  • Simple and convenient: Cells are directly cultured in 96-well plates, no cell lysis necessary
  • Accurate: Protein phosphorylation is normalized to total cellular protein in the same well
  • Method: Cell-Based ELISA (FL530/585 nm, 360/450 nm)
  • Samples: Cells
  • Species: human, rat and mouse

PROTEIN PHOSPHORYLATION plays key roles in regulating protein activity within cells. A large number of protein kinases and phosphatases are known to be involved in a variety of signal transduction pathways. A rapid assay capable of monitoring such signal transduction pathways, target protein phosphorylation status and drug effects is highly desirable in life science research and drug discovery.

Liver Kinase B1 (LKB1, STK11, PJS, hLKB1) is a member of the serine/threonine kinase family that regulates cell polarity and functions as a tumour suppressor. BioAssay Systems' LKB1(S431) Phosphorylation ELISA Kit provides a rapid and sensitive assay for measuring the phosphorylated protein pLKB1(S431) in cultured cells and normalizes the level of phosphorylation to the total protein content in the same well. This simple and efficient assay eliminates the need for cell lysate preparation and can be used to study the underlying signal pathway and effects of inhibitors, siRNA or activators on the pathway. In this assay, cells grown in a 96-well plate are fixed and permeabilized. The phosphorylation of LKB1(S431) is measured using a specific primary antibody followed by a HRP-conjugated secondary antibody with a fluorogenic substrate, and a fluorescent reagent that measures the total protein in the same well.